accessibility menu, dialog, popup

UserName

CH3HNNH2

CAS RN 18759-96-1

IUPAC Name: 3-[(methylamino)methyl]aniline
Synonyms: 3-[(methylamino)methyl]aniline
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: HBLPYXIZPMDWIO-UHFFFAOYSA-N
SMILES: CNCC1=CC(N)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results