missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-N-methylbenzamide 98.0+%, TCI America™
$80.16 - $233.00
Chemical Identifiers
| CAS | 25900-61-2 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.181 |
| MDL Number | MFCD00070630 |
| InChI Key | PYDQTASEULDNRL-UHFFFAOYSA-N |
| Synonym | 3-Aminobenzoylmethylamide |
| PubChem CID | 676526 |
| IUPAC Name | 3-amino-N-methylbenzamide |
| SMILES | CNC(=O)C1=CC(=CC=C1)N |
Chemical Identifiers
| 25900-61-2 | |
| 150.181 | |
| PYDQTASEULDNRL-UHFFFAOYSA-N | |
| 676526 | |
| CNC(=O)C1=CC(=CC=C1)N |
| C8H10N2O | |
| MFCD00070630 | |
| 3-Aminobenzoylmethylamide | |
| 3-amino-N-methylbenzamide |
Specifications
| 25900-61-2 | |
| C8H10N2O | |
| 5 g | |
| PYDQTASEULDNRL-UHFFFAOYSA-N | |
| 3-amino-N-methylbenzamide | |
| 676526 | |
| ≥98.0% (HPLC,N) | |
| 3-Amino-N-methylbenzamide |
| 119°C | |
| MFCD00070630 | |
| 3-Aminobenzoylmethylamide | |
| CNC(=O)C1=CC(=CC=C1)N | |
| 150.181 | |
| 150.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No