CAS RN 25900-61-2
CAS RN 25900-61-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Amino-N-methylbenzamide 98.0+%, TCI America™
CAS: 25900-61-2 Molecular Formula: C8H10N2O Molecular Weight (g/mol): 150.181 MDL Number: MFCD00070630 InChI Key: PYDQTASEULDNRL-UHFFFAOYSA-N Synonym: 3-Aminobenzoylmethylamide PubChem CID: 676526 IUPAC Name: 3-amino-N-methylbenzamide SMILES: CNC(=O)C1=CC(=CC=C1)N