missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methylthio-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AAB2099103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2099103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2099103 Supplier Thermo Scientific Chemicals Supplier No. B2099103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72760-85-1 | Molecular Formula: C5H6N4S | Molecular Weight: 154.191 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72760-85-1
Molecular Formula C5H6N4S
Molecular Weight (g/mol) 154.191
MDL Number MFCD00052745
InChI Key IUVUVQGOSHLPJV-UHFFFAOYSA-N
Synonym 3-amino-5-methylthio-1h-pyrazole-4-carbonitrile, 3-amino-5-methylthio pyrazole-4-carbonitrile, 3-amino-4-cyano-5-methylthiopyrazole, 5-amino-3-methylsulfanyl-1h-pyrazole-4-carbonitrile, 5-amino-3-methylthiopyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3-amino-5-methylthio, 5-amino-3-methylthio-1h-pyrazole-4-carbonitrile, 3-amino-5-methylsulfanyl-1h-pyrazole-4-carbonitrile, 3-amino-5-methylsulfanyl-2h-pyrazole-4-carbonitrile
PubChem CID 736200
IUPAC Name 5-amino-3-methylsulfanyl-1H-pyrazole-4-carbonitrile
SMILES CSC1=NNC(=C1C#N)N

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
UN Number UN3439
Formula Weight 154.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-methylthio-1H-pyrazole-4-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only