accessibility menu, dialog, popup

UserName

CAS RN 72760-85-1

IUPAC Name: 5-amino-3-(methylsulfanyl)-1H-pyrazole-4-carbonitrile
Synonyms: 3-amino-5-(methylsulfanyl)-2H-pyrazole-4-carbonitrile
Molecular Weight (g/mol): 154.19
Molecular Formula: C5H6N4S
InChi Key: IUVUVQGOSHLPJV-UHFFFAOYSA-N
SMILES: CSC1=NNC(N)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results