missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methylpyrazole 96.0+%, TCI America™

$290.70 - $290.70

Chemical Identifiers

CAS 31230-17-8
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00075180
InChI Key FYTLHYRDGXRYEY-UHFFFAOYSA-N
Synonym 3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
PubChem CID 93146
IUPAC Name 5-methyl-1H-pyrazol-3-amine
SMILES CC1=CC(=NN1)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A113325G
View Documents
TCI America
A113325G
25 g
Each for $290.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

31230-17-8
97.121
FYTLHYRDGXRYEY-UHFFFAOYSA-N
93146
CC1=CC(=NN1)N
C4H7N3
MFCD00075180
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
5-methyl-1H-pyrazol-3-amine

Specifications

31230-17-8
MFCD00075180
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
CC1=CC(=NN1)N
97.121
97.12
Crystals
C4H7N3
25 g
FYTLHYRDGXRYEY-UHFFFAOYSA-N
5-methyl-1H-pyrazol-3-amine
93146
≥96.0% (GC)
3-Amino-5-methylpyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'