missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methylpyrazole 96.0+%, TCI America™

Supplier:  TCI America A113325G

 View more versions of this product

Catalog No. A113325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Amino-5-methylpyrazole
C4H7N3
25 g
FYTLHYRDGXRYEY-UHFFFAOYSA-N
5-methyl-1H-pyrazol-3-amine
93146
≥96.0% (GC)
31230-17-8
MFCD00075180
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
CC1=CC(=NN1)N
97.121
97.12
Crystals

Chemical Identifiers

31230-17-8
97.121
FYTLHYRDGXRYEY-UHFFFAOYSA-N
93146
CC1=CC(=NN1)N
C4H7N3
MFCD00075180
3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
5-methyl-1H-pyrazol-3-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions