missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-bromo-2-chloropyridine 98.0+%, TCI America™
Supplier: TCI America A254025G
Specifications
| 3-Amino-5-bromo-2-chloropyridine | |
| 131°C | |
| C5H4BrClN2 | |
| 25 g | |
| ZSEZSALOLWCCGT-UHFFFAOYSA-N | |
| 5-bromo-2-chloropyridin-3-amine | |
| 21858354 | |
| ≥98.0% (GC) |
| 588729-99-1 | |
| White-Yellow | |
| MFCD02682092 | |
| 3-amino-5-bromo-2-chloropyridine, 2-chloro-3-amino-5-bromopyridine, 5-bromo-2-chloro-3-pyridinamine, 5-bromo-2-chloro-pyridin-3-ylamine, 5-bromo-2-chloro-3-pyridylamine, 3-pyridinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-3-aminopyridine, zlchem 394, pubchem1142, acmc-209m7r | |
| NC1=CC(Br)=CN=C1Cl | |
| 207.46 | |
| 207.46 | |
| Crystalline Powder |
Chemical Identifiers
| 588729-99-1 | |
| 207.46 | |
| ZSEZSALOLWCCGT-UHFFFAOYSA-N | |
| 21858354 | |
| NC1=CC(Br)=CN=C1Cl |
| C5H4BrClN2 | |
| MFCD02682092 | |
| 3-amino-5-bromo-2-chloropyridine, 2-chloro-3-amino-5-bromopyridine, 5-bromo-2-chloro-3-pyridinamine, 5-bromo-2-chloro-pyridin-3-ylamine, 5-bromo-2-chloro-3-pyridylamine, 3-pyridinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-3-aminopyridine, zlchem 394, pubchem1142, acmc-209m7r | |
| 5-bromo-2-chloropyridin-3-amine |
Safety and Handling
TSCA : No