missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-2-chloropyridine 98.0+%, TCI America™

Catalog No. p-7117963
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25405G 5 g
A254025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25405G Supplier TCI America Supplier No. A25405G
Only null left

Chemical Identifiers

CAS 588729-99-1
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.46
MDL Number MFCD02682092
InChI Key ZSEZSALOLWCCGT-UHFFFAOYSA-N
Synonym 3-amino-5-bromo-2-chloropyridine, 2-chloro-3-amino-5-bromopyridine, 5-bromo-2-chloro-3-pyridinamine, 5-bromo-2-chloro-pyridin-3-ylamine, 5-bromo-2-chloro-3-pyridylamine, 3-pyridinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-3-aminopyridine, zlchem 394, pubchem1142, acmc-209m7r
PubChem CID 21858354
IUPAC Name 5-bromo-2-chloropyridin-3-amine
SMILES NC1=CC(Br)=CN=C1Cl

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
Formula Weight 207.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-bromo-2-chloropyridine