missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-(methylthio)benzotrifluoride 98.0+%, TCI America™

Catalog No. a16661g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16661G Supplier TCI America Supplier No. A16661G

Chemical Identifiers

CAS 207974-07-0
Molecular Formula C8H8F3NS
Molecular Weight (g/mol) 207.214
MDL Number MFCD00052715
InChI Key OQFOLJGOVFYDHZ-UHFFFAOYSA-N
Synonym 3-amino-4-methylthio benzotrifluoride, 2-methylthio-5-trifluoromethyl aniline, 2-amino-1-methylthio-4-trifluoromethylbenzene, 2-methylsulfanyl-5-trifluoromethyl aniline, 2-amino-1-methylthio-4-triflyoromethylbenzene, 2-methylsulfanyl-5-trifluoromethyl-phenylamine, 2-methylthio-5-trifluoromethyl phenylamine, pubchem2785
PubChem CID 606105
IUPAC Name 2-methylsulfanyl-5-(trifluoromethyl)aniline
SMILES CSC1=C(C=C(C=C1)C(F)(F)F)N

Specifications

Boiling Point 112°C
Quantity 1 g
Formula Weight 207.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-4-(methylthio)benzotrifluoride