missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylphenylboronic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 455180050

 View more versions of this product

Catalog No. AC455180050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

3-Amino-4-methylphenylboronic acid
Authentic
C7H10BNO2
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
B(C1=CC(=C(C=C1)C)N)(O)O
150.98
150.98
22237-12-3
97.5% min. (HPLC)
5g
PLTGUDDQNWJILD-UHFFFAOYSA-N
(3-amino-4-methylphenyl)boronic acid
2737803
98%

Chemical Identifiers

22237-12-3
150.98
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
(3-amino-4-methylphenyl)boronic acid
C7H10BNO2
PLTGUDDQNWJILD-UHFFFAOYSA-N
2737803
B(C1=CC(=C(C=C1)C)N)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only