missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylphenol 98.0+%, TCI America™

Catalog No. p-7118244
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31801G 1 g
A31805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31801G Supplier TCI America Supplier No. A31801G

Chemical Identifiers

CAS 2836-00-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00075083
InChI Key NUNAWQZKZVVELQ-UHFFFAOYSA-N
Synonym 3-Amino-p-cresol, 5-Hydroxy-2-methylaniline
PubChem CID 17820
IUPAC Name 3-amino-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)N

Specifications

Melting Point 161°C
Quantity 1 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylphenol
EINECSNumber (3)-0707
TSCA No