accessibility menu, dialog, popup

UserName

CH3OHH2N

CAS RN 2836-00-2

IUPAC Name: 3-amino-4-methylphenol
Synonyms: phenol, 3-amino-4-methyl- P-cresol,2-amino
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: NUNAWQZKZVVELQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results