missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-methylbenzyl Alcohol 98.0+%, TCI America™
$87.60 - $260.97
Chemical Identifiers
| CAS | 81863-45-8 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.18 |
| MDL Number | MFCD00075051 |
| InChI Key | BCQKYGYTOHXGLL-UHFFFAOYSA-N |
| PubChem CID | 586243 |
| IUPAC Name | (3-amino-4-methylphenyl)methanol |
| SMILES | CC1=C(N)C=C(CO)C=C1 |
Chemical Identifiers
| 81863-45-8 | |
| 137.18 | |
| BCQKYGYTOHXGLL-UHFFFAOYSA-N | |
| (3-amino-4-methylphenyl)methanol |
| C8H11NO | |
| MFCD00075051 | |
| 586243 | |
| CC1=C(N)C=C(CO)C=C1 |
Specifications
| 81863-45-8 | |
| C8H11NO | |
| 1 g | |
| CC1=C(N)C=C(CO)C=C1 | |
| 137.18 | |
| 137.18 | |
| Crystalline Powder |
| 109°C | |
| MFCD00075051 | |
| BCQKYGYTOHXGLL-UHFFFAOYSA-N | |
| (3-amino-4-methylphenyl)methanol | |
| 586243 | |
| ≥98.0% (GC,T) | |
| 3-Amino-4-methylbenzyl Alcohol |
Safety and Handling
TSCA : No