accessibility menu, dialog, popup

UserName

CH3OHNH2

CAS RN 81863-45-8

IUPAC Name: (3-amino-4-methylphenyl)methanol
Synonyms: (3-amino-4-methylphenyl)methanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: BCQKYGYTOHXGLL-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C(CO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results