missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. A29981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29981G 1 g
A29985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29981G Supplier TCI America Supplier No. A29981G

Chemical Identifiers

CAS 81863-45-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075051
InChI Key BCQKYGYTOHXGLL-UHFFFAOYSA-N
PubChem CID 586243
IUPAC Name (3-amino-4-methylphenyl)methanol
SMILES CC1=C(N)C=C(CO)C=C1

Specifications

Melting Point 109°C
Quantity 1 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzyl Alcohol