missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7117531
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A15825G 5 g
A158225G 25 g
2 options

Catalog No. A15825G

Supplier: TCI America A15825G

Only null left

Chemical Identifiers

CAS 535-52-4
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD00007653
InChI Key DRKWGMXFFCPZLW-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl aniline, 3-amino-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-toluidine, 4-fluoro-3-aminobenzotrifluoride, 2-fluoro-5-trifluoromethyl benzenamine, 2-fluoro-5-trifluoromethyl phenylamine, benzenamine, 2-fluoro-5-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-m-toluidine, pubchem1538, ksc494s7l
PubChem CID 136349
IUPAC Name 2-fluoro-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)F

Specifications

Boiling Point 190°C
Quantity 5 g
Formula Weight 179.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Amino-4-fluorobenzotrifluoride