accessibility menu, dialog, popup

UserName

CAS RN 535-52-4

IUPAC Name: 2-fluoro-5-(trifluoromethyl)aniline
Synonyms: 2-fluoro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: DRKWGMXFFCPZLW-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1F)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)