missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-fluorobenzoic Acid 97.0+%, TCI America™

Catalog No. A19725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19725G Supplier TCI America Supplier No. A19725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2365-85-7
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00130055
InChI Key WFSPEVFSRUTRCN-UHFFFAOYSA-N
Synonym 3-amino-4-fluorobenzoicacid, benzoic acid, 3-amino-4-fluoro, 5-carboxy-2-fluoroaniline, 3-amino-4-fluoro-benzoic acid, pubchem4690, acmc-1cilf, ksc207o2p, benzoicacid, 3-amino-4-fluoro, tos-bb-0112, wfspevfsrutrcn-uhfffaoysa
PubChem CID 75396
IUPAC Name 3-amino-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)F

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-fluorobenzoic Acid