accessibility menu, dialog, popup

UserName

NH2FOHO

CAS RN 2365-85-7

IUPAC Name: 3-amino-4-fluorobenzoic acid
Synonyms: 3-amino-4-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: WFSPEVFSRUTRCN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1F)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results