missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-chlorobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America A094825G
Specifications
| 3-Amino-4-chlorobenzotrifluoride | |
| Yellow | |
| C7H5ClF3N | |
| 25 g | |
| 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda | |
| C1=CC(=C(C=C1C(F)(F)F)N)Cl | |
| 195.569 | |
| 195.57 | |
| Liquid |
| 121-50-6 | |
| 106°C | |
| MFCD00007673 | |
| 2810 | |
| VKTTYIXIDXWHKW-UHFFFAOYSA-N | |
| 2-chloro-5-(trifluoromethyl)aniline | |
| 8475 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 121-50-6 | |
| 195.569 | |
| VKTTYIXIDXWHKW-UHFFFAOYSA-N | |
| 8475 | |
| C1=CC(=C(C=C1C(F)(F)F)N)Cl |
| C7H5ClF3N | |
| MFCD00007673 | |
| 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda | |
| 2-chloro-5-(trifluoromethyl)aniline |
Safety and Handling
EINECSNumber : (3)-0266
RTECSNumber : XU5775000
TSCA : Yes