accessibility menu, dialog, popup

UserName

CAS RN 121-50-6

IUPAC Name: 2-chloro-5-(trifluoromethyl)aniline
Synonyms: 2-chloro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: VKTTYIXIDXWHKW-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)