missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. A0948500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0948500G 500 g
A094825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0948500G Supplier TCI America Supplier No. A0948500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-50-6
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.569
MDL Number MFCD00007673
InChI Key VKTTYIXIDXWHKW-UHFFFAOYSA-N
Synonym 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda
PubChem CID 8475
IUPAC Name 2-chloro-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)Cl

Specifications

Color Yellow
Boiling Point 106°C
Quantity 500 g
UN Number 2810
Formula Weight 195.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Amino-4-chlorobenzotrifluoride
EINECSNumber (3)-0266
RTECSNumber XU5775000
TSCA Yes