missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7116980
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A063525G 25 g
A0635500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A063525G Supplier TCI America Supplier No. A063525G
Only null left

Chemical Identifiers

CAS 2840-28-0
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007671
InChI Key DMGFVJVLVZOSOE-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213
PubChem CID 76092
IUPAC Name 3-amino-4-chlorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)Cl

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 25 g
Formula Weight 171.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-chlorobenzoic Acid
EINECSNumber (3)-1480
TSCA Yes