accessibility menu, dialog, popup

UserName

CAS RN 2840-28-0

IUPAC Name: 3-amino-4-chlorobenzoic acid
Synonyms: 3-amino-4-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: DMGFVJVLVZOSOE-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)C(O)=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)