missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117908
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24415G 5 g
A244125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24415G Supplier TCI America Supplier No. A24415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2840-29-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00579106
InChI Key TZFJADFQEBASQU-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-benzoic acid, benzoic acid, 3-amino-4-bromo, 3-amino-4-bromobenzoicacid, acmc-1cpid, 4-brom-3-aminobenzoesaure, ksc494k6h, 3-amino-4-bromobenzoic acid, 3-amino-4-bromo benzoic acid
PubChem CID 3418752
IUPAC Name 3-amino-4-bromobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)Br

Specifications

Melting Point 228°C
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromobenzoic Acid