accessibility menu, dialog, popup

UserName

CAS RN 2840-29-1

IUPAC Name: 3-amino-4-bromobenzoic acid
Synonyms: 3-amino-4-bromobenzoic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: TZFJADFQEBASQU-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=CC(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results