missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylbenzoic Acid 99.0+%, TCI America™

Catalog No. A140125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A140125G 25 g
A1401250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A140125G Supplier TCI America Supplier No. A140125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52130-17-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00075026
InChI Key BYHMLZGICSEKIY-UHFFFAOYSA-N
Synonym 2-methyl-3-aminobenzoic acid, 3-amino-o-toluic acid, 3-amino-2-methylbenzoicacid, 3-amino-2-methyl-benzoic acid, benzoic acid, 3-amino-2-methyl, 2-methyl-3-amino benzoic acid, pubchem10921, acmc-209ky1, ksc489o4b, 3-amino-2-methyl benzoic acid
PubChem CID 2733699
IUPAC Name 3-amino-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1N)C(=O)O

Specifications

Melting Point 188°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-methylbenzoic Acid
EINECSNumber (3)-4438
TSCA No