accessibility menu, dialog, popup

UserName

CAS RN 52130-17-3

IUPAC Name: 3-amino-2-methylbenzoate
Synonyms: 3-amino-2-methylbenzoate
Molecular Weight (g/mol): 150.16
Molecular Formula: C8H8NO2
InChi Key: BYHMLZGICSEKIY-UHFFFAOYSA-M
SMILES: CC1=C(N)C=CC=C1C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)