missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. A29381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29381G 1 g
A29385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29381G Supplier TCI America Supplier No. A29381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 861306-04-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09701221
InChI Key ZBQFWVIYNLHFMO-UHFFFAOYSA-N
Synonym 3-Amino-o-anisic Acid
PubChem CID 19849158
IUPAC Name 3-amino-2-methoxybenzoic acid
SMILES COC1=C(C=CC=C1N)C(=O)O

Specifications

Melting Point 101°C
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-methoxybenzoic Acid