missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,8-Dibromo-1,10-phenanthroline 96.0+%, TCI America™
Supplier: TCI America D32091G
Specifications
| 3,8-Dibromo-1,10-phenanthroline | |
| 282°C | |
| C12H6Br2N2 | |
| 1 g | |
| BrC1=CN=C2C(C=CC3=CC(Br)=CN=C23)=C1 | |
| 338.00 | |
| 338.00 | |
| Crystalline Powder |
| 100125-12-0 | |
| White-Yellow | |
| MFCD09909860 | |
| IDWJREBUVYSPKS-UHFFFAOYSA-N | |
| 3,8-dibromo-1,10-phenanthroline | |
| 10991348 | |
| ≥96.0% (HPLC,T) |
Chemical Identifiers
| 100125-12-0 | |
| 338.00 | |
| IDWJREBUVYSPKS-UHFFFAOYSA-N | |
| 3,8-dibromo-1,10-phenanthroline |
| C12H6Br2N2 | |
| MFCD09909860 | |
| 10991348 | |
| BrC1=CN=C2C(C=CC3=CC(Br)=CN=C23)=C1 |
Safety and Handling
TSCA : No