accessibility menu, dialog, popup

UserName

NNBrBr

CAS RN 100125-12-0

IUPAC Name: 3,8-dibromo-1,10-phenanthroline
Synonyms: 3,8-dibromo-1,10-phenanthroline
Molecular Weight (g/mol): 338
Molecular Formula: C12H6Br2N2
InChi Key: IDWJREBUVYSPKS-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C(C=CC3=CC(Br)=CN=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results