missing translation for 'onlineSavingsMsg'
Learn More

3,8-Dibromo-1,10-phenanthroline 96.0+%, TCI America™

Catalog No. p-7127623
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3209200MG 200 mg
D32091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3209200MG Supplier TCI America Supplier No. D3209200MG

Chemical Identifiers

CAS 100125-12-0
Molecular Formula C12H6Br2N2
Molecular Weight (g/mol) 338.00
MDL Number MFCD09909860
InChI Key IDWJREBUVYSPKS-UHFFFAOYSA-N
PubChem CID 10991348
IUPAC Name 3,8-dibromo-1,10-phenanthroline
SMILES BrC1=CN=C2C(C=CC3=CC(Br)=CN=C23)=C1

Specifications

Melting Point 282°C
Color White-Yellow
Quantity 200 mg
Formula Weight 338.00
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,8-Dibromo-1,10-phenanthroline