missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dithia-1,9-nonanedithiol 97.0+%, TCI America™

Catalog No. D28525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28525G 5 g
D28521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28525G Supplier TCI America Supplier No. D28525G
Only null left

Chemical Identifiers

CAS 25676-62-4
Molecular Formula C7H16S4
Molecular Weight (g/mol) 228.45
MDL Number MFCD00506918
InChI Key XYHKMRNYSMEEDT-UHFFFAOYSA-N
PubChem CID 373740
IUPAC Name 2-({3-[(2-sulfanylethyl)sulfanyl]propyl}sulfanyl)ethane-1-thiol
SMILES SCCSCCCSCCS

Specifications

Color Yellow
Quantity 5 g
Synonym 1,3-Bis(2-mercaptoethylthio)propane, 2,2′-(Trimethylenedithio)diethanethiol
Formula Weight 228.45
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,7-Dithia-1,9-nonanedithiol