accessibility menu, dialog, popup

UserName

SHSSSH

CAS RN 25676-62-4

IUPAC Name: 2-({3-[(2-sulfanylethyl)sulfanyl]propyl}sulfanyl)ethane-1-thiol
Synonyms: 2-({3-[(2-sulfanylethyl)sulfanyl]propyl}sulfanyl)ethanethiol
Molecular Weight (g/mol): 228.45
Molecular Formula: C7H16S4
InChi Key: XYHKMRNYSMEEDT-UHFFFAOYSA-N
SMILES: SCCSCCCSCCS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results