missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dithia-1,8-octanediol 98.0+%, TCI America™

Catalog No. p-7127159
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D218825G 25 g
D2188250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D218825G Supplier TCI America Supplier No. D218825G
Only null left

Chemical Identifiers

CAS 5244-34-8
Molecular Formula C6H14O2S2
Molecular Weight (g/mol) 182.296
MDL Number MFCD00002911
InChI Key PDHFSBXFZGYBIP-UHFFFAOYSA-N
Synonym 3,6-dithia-1,8-octanediol, 1,2-bis 2-hydroxyethylthio ethane, 3,6-dithiaoctane-1,8-diol, dithiaoctanediol, ethylenedithioethanol, ethanol, 2,2'-1,2-ethanediylbis thio bis, unii-kbc80263g6, 2,2'-ethane-1,2-diylbis sulfanediyl diethanol, 2,2'-ethylenedithio diethanol, ethanol, 2,2'-ethylenedithio di
PubChem CID 78904
IUPAC Name 2-[2-(2-hydroxyethylsulfanyl)ethylsulfanyl]ethanol
SMILES C(CSCCSCCO)O

Specifications

Melting Point 65°C
Color White
Boiling Point 100°C
Quantity 25 g
Formula Weight 182.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dithia-1,8-octanediol
EINECSNumber (2)-3123
TSCA Yes