missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dimethyl-4-octyne-3,6-diol 98.0+%, TCI America™
Supplier: TCI America D116825G
Specifications
| 3,6-Dimethyl-4-octyne-3,6-diol | |
| 54°C | |
| 135°C | |
| MFCD00004480 | |
| 3,6-Dihydroxy-3,6-dimethyl-4-octyne | |
| CCC(C)(C#CC(C)(CC)O)O | |
| 170.252 | |
| 170.25 | |
| Crystalline Powder |
| 78-66-0 | |
| White | |
| C10H18O2 | |
| 25 g | |
| NUYADIDKTLPDGG-UHFFFAOYSA-N | |
| 3,6-dimethyloct-4-yne-3,6-diol | |
| 6546 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 78-66-0 | |
| 170.252 | |
| NUYADIDKTLPDGG-UHFFFAOYSA-N | |
| 6546 | |
| CCC(C)(C#CC(C)(CC)O)O |
| C10H18O2 | |
| MFCD00004480 | |
| 3,6-Dihydroxy-3,6-dimethyl-4-octyne | |
| 3,6-dimethyloct-4-yne-3,6-diol |
Safety and Handling
EINECSNumber : (2)-0269
RTECSNumber : RI2650000
TSCA : Yes