accessibility menu, dialog, popup

UserName

CH3CH3OHCH3OHH3C

CAS RN 78-66-0

IUPAC Name: 3,6-dimethyloct-4-yne-3,6-diol
Synonyms: 3,6-dimethyloct-4-yne-3,6-diol
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: NUYADIDKTLPDGG-UHFFFAOYNA-N
SMILES: CCC(C)(O)C#CC(C)(O)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results