missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diiodocarbazole 98.0+%, TCI America™
$206.90 - $671.06
Chemical Identifiers
| CAS | 57103-02-3 |
|---|---|
| Molecular Formula | C12H7I2N |
| Molecular Weight (g/mol) | 419.00 |
| MDL Number | MFCD00450017 |
| InChI Key | PECAOKZHORDWAI-UHFFFAOYSA-N |
| PubChem CID | 3116488 |
| IUPAC Name | 3,6-diiodo-9H-carbazole |
| SMILES | IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1 |
Chemical Identifiers
| 57103-02-3 | |
| 419.00 | |
| PECAOKZHORDWAI-UHFFFAOYSA-N | |
| 3,6-diiodo-9H-carbazole |
| C12H7I2N | |
| MFCD00450017 | |
| 3116488 | |
| IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1 |
Specifications
| 57103-02-3 | |
| C12H7I2N | |
| 5 g | |
| IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1 | |
| 419.00 | |
| 419.00 | |
| Crystalline Powder |
| 207°C | |
| MFCD00450017 | |
| PECAOKZHORDWAI-UHFFFAOYSA-N | |
| 3,6-diiodo-9H-carbazole | |
| 3116488 | |
| ≥98.0% (GC) | |
| 3,6-Diiodocarbazole |
Safety and Handling
TSCA : No