accessibility menu, dialog, popup

UserName

INHI

CAS RN 57103-02-3

IUPAC Name: 3,6-diiodo-9H-carbazole
Synonyms: 3,6-diiodo-9H-carbazole
Molecular Weight (g/mol): 419
Molecular Formula: C12H7I2N
InChi Key: PECAOKZHORDWAI-UHFFFAOYSA-N
SMILES: IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results