missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diiodocarbazole 98.0+%, TCI America™
Supplier: TCI America D454825G
Specifications
| 3,6-Diiodocarbazole | |
| 207°C | |
| MFCD00450017 | |
| PECAOKZHORDWAI-UHFFFAOYSA-N | |
| 3,6-diiodo-9H-carbazole | |
| 3116488 | |
| ≥98.0% (GC) |
| 57103-02-3 | |
| C12H7I2N | |
| 25 g | |
| IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1 | |
| 419.00 | |
| 419.00 | |
| Crystalline Powder |
Chemical Identifiers
| 57103-02-3 | |
| 419.00 | |
| PECAOKZHORDWAI-UHFFFAOYSA-N | |
| 3,6-diiodo-9H-carbazole |
| C12H7I2N | |
| MFCD00450017 | |
| 3116488 | |
| IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1 |
Safety and Handling
TSCA : No