missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydroxynaphthalene-2,7-disulfonic Acid Disodium Salt 98.0+%, TCI America™

Catalog No. p-7126310
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059525G 25 g
D0595500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D059525G Supplier TCI America Supplier No. D059525G

Chemical Identifiers

CAS 7153-21-1
Molecular Formula C10H8NaO8S2
Molecular Weight (g/mol) 343.28
MDL Number MFCD00004090
InChI Key WZCUTQZDUZZQSZ-UHFFFAOYSA-N
Synonym disodium 3,6-dihydroxynaphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 3,6-dihydroxy-, disodium salt, disodium 3,6-dihydroxynaphthalene-2,7-disulphonate, 2,7-dihydroxynaphthalene-3,6-disulfonic acid disodium salt, 3,6-dihydroxynaphthalene-2,7-disulfonic acid disodium salt, 2,7-naphthalenedisulfonic acid, 3,6-dihydroxy-, sodium salt 1:2, sodium 2,7-dioxynaphthalene-3,6-disulfonate, 3,6-dihydroxy-2,7-naphthalenedisulfonic acid, disodium salt, sodium 3,6-dihydroxynaphthalene-2,7-disulfonate, acmc-20al4l
PubChem CID 81587
IUPAC Name 3,6-dihydroxynaphthalene-2,7-disulfonic acid sodium
SMILES [Na].OC1=CC2=CC(O)=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O

Specifications

Quantity 25 g
Formula Weight 364.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dihydroxynaphthalene-2,7-disulfonic Acid Disodium Salt
EINECSNumber (4)-1095
TSCA Yes