accessibility menu, dialog, popup

UserName

NaHOOHSHOOOSOHOO

CAS RN 7153-21-1

IUPAC Name: 3,6-dihydroxynaphthalene-2,7-disulfonic acid sodium
Synonyms: 3,6-dihydroxynaphthalene-2,7-disulfonic acid sodium
Molecular Weight (g/mol): 343.28
Molecular Formula: C10H8NaO8S2
InChi Key: WZCUTQZDUZZQSZ-UHFFFAOYSA-N
SMILES: [Na].OC1=CC2=CC(O)=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results