missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromothieno[3,2-b]thiophene 98.0+%, TCI America™
Supplier: TCI America D3679200MG
Specifications
| 3,6-Dibromothieno[3,2-b]thiophene | |
| 127°C | |
| C6H2Br2S2 | |
| 200 mg | |
| BrC1=CSC2=C1SC=C2Br | |
| 298.01 | |
| 298.01 | |
| Crystalline Powder |
| 392662-65-6 | |
| White-Yellow | |
| MFCD10000958 | |
| DFMPVYZMICAYCS-UHFFFAOYSA-N | |
| 3,6-dibromothieno[3,2-b]thiophene | |
| 10613979 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 392662-65-6 | |
| 298.01 | |
| DFMPVYZMICAYCS-UHFFFAOYSA-N | |
| 3,6-dibromothieno[3,2-b]thiophene |
| C6H2Br2S2 | |
| MFCD10000958 | |
| 10613979 | |
| BrC1=CSC2=C1SC=C2Br |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator