missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromothieno[3,2-b]thiophene 98.0+%, TCI America™

Catalog No. p-7127859
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3679200MG 200 mg
D36791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3679200MG Supplier TCI America Supplier No. D3679200MG
Only null left

Chemical Identifiers

CAS 392662-65-6
Molecular Formula C6H2Br2S2
Molecular Weight (g/mol) 298.01
MDL Number MFCD10000958
InChI Key DFMPVYZMICAYCS-UHFFFAOYSA-N
PubChem CID 10613979
IUPAC Name 3,6-dibromothieno[3,2-b]thiophene
SMILES BrC1=CSC2=C1SC=C2Br

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 200 mg
Formula Weight 298.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromothieno[3,2-b]thiophene
TSCA No
Recommended Storage Refrigerator