accessibility menu, dialog, popup

UserName

SSBrBr

CAS RN 392662-65-6

IUPAC Name: 3,6-dibromothieno[3,2-b]thiophene
Synonyms: 3,6-dibromothieno[3,2-b]thiophene
Molecular Weight (g/mol): 298.01
Molecular Formula: C6H2Br2S2
InChi Key: DFMPVYZMICAYCS-UHFFFAOYSA-N
SMILES: BrC1=CSC2=C1SC=C2Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results