missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-2,7-dihydroxynaphthalene 98.0+%, TCI America™

Catalog No. D46695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46695G 5 g
D46691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46695G Supplier TCI America Supplier No. D46695G

Chemical Identifiers

CAS 96965-79-6
Molecular Formula C10H6Br2O2
Molecular Weight (g/mol) 317.964
MDL Number MFCD00277643
InChI Key UAECXNARJOIHAO-UHFFFAOYSA-N
Synonym 3,6-Dibromo-2,7-naphthalenediol
PubChem CID 2795148
IUPAC Name 3,6-dibromonaphthalene-2,7-diol
SMILES C1=C2C=C(C(=CC2=CC(=C1O)Br)Br)O

Specifications

Melting Point 188°C
Quantity 5 g
Formula Weight 317.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-2,7-dihydroxynaphthalene