accessibility menu, dialog, popup

UserName

HOBrBrOH

CAS RN 96965-79-6

IUPAC Name: 3,6-dibromonaphthalene-2,7-diol
Synonyms: 3,6-dibromonaphthalene-2,7-diol
Molecular Weight (g/mol): 317.96
Molecular Formula: C10H6Br2O2
InChi Key: UAECXNARJOIHAO-UHFFFAOYSA-N
SMILES: OC1=CC2=CC(O)=C(Br)C=C2C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results