missing translation for 'onlineSavingsMsg'
Learn More

3,6-Bis(hydroxymethyl)durene 99.0+%, TCI America™

Catalog No. B04935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04935G 5 g
B04931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04935G Supplier TCI America Supplier No. B04935G
Only null left

Chemical Identifiers

CAS 7522-62-5
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00004623
InChI Key KDJOOHBQJRVMIX-UHFFFAOYSA-N
Synonym 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha′C-diol
PubChem CID 82026
IUPAC Name [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol
SMILES CC1=C(C)C(CO)=C(C)C(C)=C1CO

Specifications

Melting Point 256°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Bis(hydroxymethyl)durene