accessibility menu, dialog, popup

UserName

CAS RN 7522-62-5

CH3CH3OHCH3H3CHO

CAS RN 7522-62-5

IUPAC Name: [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol
Synonyms: [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: KDJOOHBQJRVMIX-UHFFFAOYSA-N
SMILES: CC1=C(C)C(CO)=C(C)C(C)=C1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results